Gene Transcripts

Gene Symbol
PSG3
Entrez Gene ID
5671
Gene Name
pregnancy specific beta-1-glycoprotein 3
Gene Aliases
Chromosome Location
Chr. 19: 42721642 - 42740516 on Build GRCh38
UniGene ID
Hs.654413
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021016.3 NP_066296.2 7 1637
XM_011527126.1 XP_011525428.1 6 1358
XM_011527127.1 XP_011525429.1 5 1103
GenBank mRNA BC005924.1 - 7 1602
M23575.1 - 7 1734
M27907.1 - 4 720
M30628.1 - 9 1665
M34420.1 - 7 1586
M37399.1 - 7 1589
M93061.1 - 7 1563
S59493.1 - 7 1563
S59494.1 - 9 1671

View Full Product Details