Gene Transcripts

Gene Symbol
LARP6
Entrez Gene ID
55323
Gene Name
La ribonucleoprotein domain family member 6
Gene Aliases
ACHN
Chromosome Location
Chr. 15: 70831524 - 70854265 on Build GRCh38
UniGene ID
Hs.416755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018357.3 NP_060827.2 1 320
NM_197958.2 NP_932062.1 1 320
GenBank mRNA AF443828.1 - 1 213
AF443829.1 - 1 213
AK002058.1 - 1 214
AL833876.1 - 1 174
BC006082.1 - 1 167 2961587
BC009446.1 - 1 167 3529516
BC014018.2 - 1 171 4127153
BC039153.1 - 1 176
BI913258.1 - 1 228
DN992782.1 - 1 325

View Full Product Details