Gene Transcripts
- Gene Symbol
- MAGEA5
- Entrez Gene ID
- 4104
- Gene Name
- MAGE family member A5
- Gene Aliases
- CT1.5, MAGE5, MAGEA4
- Chromosome Location
- Chr. X: 152114049 - 152117939 on Build GRCh38
- UniGene ID
- Hs.18048
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021049.4 |
NP_066387.1
|
3 |
868 |
|
- Gene Symbol
- MAGEA10-MAGEA5
- Entrez Gene ID
- 100533997
- Gene Name
- MAGEA10-MAGEA5 readthrough
- Gene Aliases
- CT1.5
- Chromosome Location
- Chr. X: 152114049 - 152138578 on Build GRCh38
- UniGene ID
- Hs.18048
- Species
- Homo sapiens
View Full Product Details