Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001308419.1 | NP_001295348.1 | 5 | 461 | |
NM_022165.2 | NP_071448.1 | 6 | 671 | ||
XM_006723323.3 | XP_006723386.1 | 5 | 619 | ||
XM_017027131.1 | XP_016882620.1 | 4 | 863 | ||
GenBank mRNA | AF311862.1 | - | 6 | 649 | |
AY358744.1 | - | 6 | 671 | ||
BC027618.2 | - | 6 | 645 | 4830017 | |
BM069859.1 | - | 5 | 419 |