Gene Transcripts

Gene Symbol
MSH3
Entrez Gene ID
4437
Gene Name
mutS homolog 3
Gene Aliases
DUP, MRP1
Chromosome Location
Chr. 5: 80654648 - 80876815 on Build GRCh38
UniGene ID
Hs.280987
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002439.4 NP_002430.3 3, 4 667
GenBank mRNA AK289856.1 - 4, 5 636
BC004177.2 - 4, 5 608
BC011817.2 - 4, 5 608
BC017273.2 - 3, 4 590
BC098436.1 - 4, 5 608
BC130434.1 - 3, 4 667 40146465
BC130436.1 - 3, 4 667 40146467
J04810.1 - 3, 4 648
U61981.1 - 4, 5 576

View Full Product Details