Gene Transcripts

Gene Symbol
DGCR8
Entrez Gene ID
54487
Gene Name
DGCR8 microprocessor complex subunit
Gene Aliases
C22orf12, DGCRK6, Gy1, pasha
Chromosome Location
Chr. 22: 20080232 - 20111877 on Build GRCh38
UniGene ID
Hs.643452
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190326.1 NP_001177255.1 2 669
NM_022720.6 NP_073557.3 2 669
GenBank mRNA AF165527.1 - 1 330
AK313357.1 - 2 620
BC009984.2 - 2 590
BC037564.1 - 2 619
BC078147.1 - 1 383 6014346
BQ227899.1 - 2 626
CR456356.1 - 1 327

View Full Product Details