Gene Transcripts

Gene Symbol
GINS3
Entrez Gene ID
64785
Gene Name
GINS complex subunit 3
Gene Aliases
PSF3
Chromosome Location
Chr. 16: 58392394 - 58406144 on Build GRCh38
UniGene ID
Hs.47125
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126129.1 NP_001119601.1 2, 3 519
NM_022770.3 NP_073607.2 1, 2 402
GenBank mRNA AK127454.1 - 1 164
AK315622.1 - 1, 2 353
BC005879.1 - 1, 2 277 3529058
BC014437.1 - 1, 2 296 4870865
BP374281.1 - 2, 3 462
DA550725.1 - 1, 2 402
KU178752.1 - 1, 2 194
KU178753.1 - 2, 3 311

View Full Product Details