Gene Transcripts

Gene Symbol
SHCBP1
Entrez Gene ID
79801
Gene Name
SHC binding and spindle associated 1
Gene Aliases
PAL
Chromosome Location
Chr. 16: 46578591 - 46624933 on Build GRCh38
UniGene ID
Hs.123253
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324318.1 NP_001311247.1 1, 2 154
NM_001324319.1 NP_001311248.1 1, 2 154
NM_024745.4 NP_079021.3 1, 2 151
NR_136738.1 1, 2 154
XM_011523335.2 XP_011521637.1 1, 2 3685
GenBank mRNA AK055931.1 - 1, 2 122
AK292164.1 - 1, 2 118
AK310430.1 - 1, 2 120
AK315706.1 - 1, 2 120
BC030699.1 - 1, 2 126 4829528
BP245197.1 - 1, 2 151

View Full Product Details