Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195263.1 | NP_001182192.1 | 2 | 392 | |
NM_024895.4 | NP_079171.1 | 2 | 392 | ||
XM_005270165.2 | XP_005270222.1 | 2 | 367 | ||
XM_011540177.2 | XP_011538479.1 | 3 | 552 | ||
XM_011540178.2 | XP_011538480.1 | 2 | 367 | ||
XM_011540179.2 | XP_011538481.1 | 2 | 367 | ||
XM_011540181.2 | XP_011538483.1 | 2 | 367 | ||
XM_011540182.2 | XP_011538484.1 | 2 | 367 | ||
XM_011540183.2 | XP_011538485.1 | 2 | 367 | ||
XM_017016667.1 | XP_016872156.1 | 2 | 367 | ||
XM_017016668.1 | XP_016872157.1 | 2 | 367 | ||
GenBank mRNA | AK024422.1 | - | 2 | 340 | |
AK026862.1 | - | 2 | 357 | ||
AK225136.1 | - | 2 | 364 | ||
BC029054.1 | - | 1 | 317 | 5179973 | |
DA197280.1 | - | 2 | 393 | ||
FJ617449.1 | - | 3 | 270 |