Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030651.3 | NP_085154.3 | 4 | 1610 | |
GenBank mRNA | AF085841.1 | - | 1 | 267 | |
AK054885.1 | - | 4 | 1610 | ||
AK123424.1 | - | 3 | 2492 | ||
BC013201.1 | - | 6 | 1436 | 3924575 | |
BC063046.1 | - | 4 | 1558 | 5197474 |