Gene Transcripts

Gene Symbol
RNF32
Entrez Gene ID
140545
Gene Name
ring finger protein 32
Gene Aliases
FKSG33, HSD15, LMBR2
Chromosome Location
Chr. 7: 156640286 - 156678333 on Build GRCh38
UniGene ID
Hs.446194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184996.1 NP_001171925.1 6 805
NM_001184997.1 NP_001171926.1 6 658
NM_001308273.1 NP_001295202.1 6 756
NM_001308274.1 NP_001295203.1 5 547
NM_030936.3 NP_112198.1 6 700
NR_131778.1 6 654
XM_005249522.4 XP_005249579.1 6 612
XM_011515804.2 XP_011514106.1 7 722
XM_011515805.2 XP_011514107.1 7 879
XM_011515806.2 XP_011514108.1 6 790
XM_011515807.2 XP_011514109.1 6 598
XM_011515808.2 XP_011514110.1 6 710
XM_011515809.2 XP_011514111.1 6 783
XM_011515810.2 XP_011514112.1 6 600
XM_011515811.2 XP_011514113.1 6 595
XM_011515812.2 XP_011514114.1 6 702
XM_017011753.1 XP_016867242.1 6 704
XM_017011754.1 XP_016867243.1 6 704
XM_017011755.1 XP_016867244.1 6 704
GenBank mRNA AF325690.1 - 6 608
AF441222.1 - 5 547
AF441224.1 - 6 654
AK307316.1 - 7 772
AK314067.1 - 6 805
AL136874.1 - 6 588
BC015416.1 - 5 558 4395421
BT007037.1 - 4 470
CR533513.1 - 4 470

View Full Product Details