Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_182663.3 | NP_872604.1 | 3 | 708 | |
NM_182664.3 | NP_872605.1 | 3 | 708 | ||
NM_182665.3 | NP_872606.1 | 2 | 586 | ||
GenBank mRNA | AB209145.1 | - | 3 | 643 | |
AF445801.1 | - | 2 | 586 | ||
AK074115.1 | - | 1 | 53 | ||
AK289861.1 | - | 3 | 708 | ||
AK291268.1 | - | 3 | 706 | ||
AK292039.1 | - | 3 | 444 | ||
AL832784.1 | - | 2 | 486 | ||
AY062002.1 | - | 3 | 692 | ||
AY062003.1 | - | 3 | 692 | ||
AY216268.1 | - | 2 | 579 | ||
AY261332.1 | - | 2 | 486 | ||
BC004270.1 | - | 3 | 692 | 3613993 | |
BC007203.1 | - | 3 | 692 | 2967057 | |
BC042651.1 | - | 2 | 566 | 4342962 |