Gene Transcripts

Gene Symbol
MFSD7
Entrez Gene ID
84179
Gene Name
major facilitator superfamily domain containing 7
Gene Aliases
LP2561
Chromosome Location
Chr. 4: 676205 - 689468 on Build GRCh38
UniGene ID
Hs.567612
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001294341.1 NP_001281270.1 6 1063
NM_001294342.1 NP_001281271.1 4 706
NM_032219.3 NP_115595.2 6 1060
XM_005272297.1 XP_005272354.1 4 796
XM_005272299.4 XP_005272356.1 6 1090
XM_006713918.1 XP_006713981.1 6 717
XM_006713919.1 XP_006713982.1 4 799
XM_011513568.1 XP_011511870.1 6 1867
XM_011513571.1 XP_011511873.1 7 887
XM_011513572.1 XP_011511874.1 4 883
XM_011513573.2 XP_011511875.1 6 1090
XM_011513574.1 XP_011511876.1 5 856
XM_011513575.2 XP_011511877.1 5 1635
GenBank mRNA AK025922.1 - 4 538
AK292010.1 - 6 958
AY203936.1 - 2 409
AY358401.1 - 6 765
BC030246.1 - 6 803 5216645

View Full Product Details