Gene Transcripts
- Gene Symbol
- RAB6C
- Entrez Gene ID
- 84084
- Gene Name
- RAB6C, member RAS oncogene family
- Gene Aliases
- WTH3
- Chromosome Location
- Chr. 2: 129979662 - 129982738 on Build GRCh38
- UniGene ID
- Hs.591552
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL136727.1 |
- |
1 |
2277 |
|
View Full Product Details