Gene Transcripts

Gene Symbol
IFT43
Entrez Gene ID
112752
Gene Name
intraflagellar transport 43
Gene Aliases
C14orf179, CED3
Chromosome Location
Chr. 14: 75985753 - 76084073 on Build GRCh38
UniGene ID
Hs.532626
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102564.1 NP_001096034.1 2 138
NM_001255995.1 NP_001242924.1 2 138
NM_052873.2 NP_443105.2 2 138
NR_045664.1 2 138
NR_045665.1 2 138
GenBank mRNA AK056735.1 - 2 111
AK302944.1 - 2 121
BC010436.1 - 2 109 3608220
BI821833.1 - 2 114
CB853974.1 - 2 99
DB443802.1 - 2 138
DB497730.1 - 2 121

View Full Product Details