Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001174156.1 | NP_001167627.1 | 2 | 184 | |
NM_144660.2 | NP_653261.1 | 2 | 240 | ||
XM_005269541.4 | XP_005269598.1 | 2 | 375 | ||
XM_011539311.1 | XP_011537613.1 | 2 | 303 | ||
XM_011539312.2 | XP_011537614.1 | 2 | 233 | ||
XM_017015738.1 | XP_016871227.1 | 2 | 240 | ||
XM_017015739.1 | XP_016871228.1 | 2 | 268 | ||
GenBank mRNA | AK057811.1 | - | 2 | 168 | |
BC080593.1 | - | 2 | 184 | ||
EU668332.1 | - | 2 | 168 | ||
GQ472208.1 | - | 2 | 168 |