Gene Transcripts

Gene Symbol
TCEB3C
Entrez Gene ID
162699
Gene Name
transcription elongation factor B subunit 3C
Gene Aliases
HsT829, TCEB3L2
Chromosome Location
Chr. 18: 47028202 - 47030078 on Build GRCh38
UniGene ID
Hs.515381
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145653.3 NP_663628.2 1 1425
GenBank mRNA AB076840.1 - 1 1189

Gene Symbol
LOC107983955
Entrez Gene ID
107983955
Gene Name
RNA polymerase II transcription factor SIII subunit A3-like-1
Gene Aliases
Chromosome Location
Chr. 18: 46968565 - 46969909 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017026108.1 XP_016881597.1 1 763

Gene Symbol
TCEB3CL
Entrez Gene ID
728929
Gene Name
transcription elongation factor B subunit 3C like
Gene Aliases
HsT828, eloA3-like-1
Chromosome Location
Chr. 18: 47022287 - 47023927 on Build GRCh38
UniGene ID
Hs.745535
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100817.1 NP_001094287.1 2 1189
GenBank mRNA BC103912.1 - 2 1185
BC103913.1 - 2 1190 40006027

Gene Symbol
TCEB3CL2
Entrez Gene ID
100506888
Gene Name
transcription elongation factor B subunit 3C like 2
Gene Aliases
Chromosome Location
Chr. 18: 46962768 - 46964644 on Build GRCh38
UniGene ID
Hs.729823
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242907.1 NP_001229836.1 1 1425

View Full Product Details