Gene Transcripts

Gene Symbol
LACE1
Entrez Gene ID
246269
Gene Name
lactation elevated 1
Gene Aliases
AFG1, c222389
Chromosome Location
Chr. 6: 108294874 - 108525784 on Build GRCh38
UniGene ID
Hs.259666
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323005.1 NP_001309934.1 2, 3 563
NM_145315.4 NP_660358.2 2, 3 563
XM_005266885.3 XP_005266942.1 3, 4 1921
XM_011535657.2 XP_011533959.1 2, 3 446
XM_011535659.2 XP_011533961.1 2, 3 446
XM_011535660.2 XP_011533962.1 2, 3 446
XM_011535661.2 XP_011533963.1 2, 3 446
XM_017010647.1 XP_016866136.1 3, 4 413
GenBank mRNA AF520418.1 - 2, 3 543
BC018445.1 - 2, 3 543 3871205
BP246728.1 - 570

View Full Product Details