Gene Transcripts

Gene Symbol
SPATA2L
Entrez Gene ID
124044
Gene Name
spermatogenesis associated 2 like
Gene Aliases
C16orf76, tamo
Chromosome Location
Chr. 16: 89696357 - 89701993 on Build GRCh38
UniGene ID
Hs.374556
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152339.3 NP_689552.2 2 376
XM_005256279.4 XP_005256336.1 2 793
GenBank mRNA AK127372.1 - 1 180
BC027606.1 - 2 373
BC039608.1 - 2 354 5248678
BC053588.1 - 2 358 5245683
BI917028.1 - 2 358

View Full Product Details