Gene Transcripts
- Gene Symbol
- POC5
- Entrez Gene ID
- 134359
- Gene Name
- POC5 centriolar protein
- Gene Aliases
- C5orf37
- Chromosome Location
- Chr. 5: 75674198 - 75717488 on Build GRCh38
- UniGene ID
- Hs.432726
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001099271.1 |
NP_001092741.1
|
7, 8 |
997 |
|
NM_152408.2 |
NP_689621.2
|
6, 7 |
796 |
|
XM_005248436.1 |
XP_005248493.1
|
7, 8 |
997 |
|
XM_011543158.2 |
XP_011541460.1
|
7, 8 |
997 |
|
XM_011543159.1 |
XP_011541461.1
|
8, 9 |
1003 |
|
XM_011543160.1 |
XP_011541462.1
|
6, 7 |
857 |
|
XM_017009038.1 |
XP_016864527.1
|
6, 7 |
917 |
|
XM_017009039.1 |
XP_016864528.1
|
6, 7 |
836 |
|
GenBank mRNA |
AK093098.1 |
- |
6, 7 |
766 |
|
AK296071.1 |
- |
7, 8 |
914 |
|
BC101325.1 |
- |
6, 7 |
762 |
40024984 |
BC101326.2 |
- |
6, 7 |
766 |
40024987 |
BC101327.2 |
- |
6, 7 |
766 |
40024989 |
BC101328.2 |
- |
6, 7 |
763 |
|
View Full Product Details