Gene Transcripts

Gene Symbol
OIT3
Entrez Gene ID
170392
Gene Name
oncoprotein induced transcript 3
Gene Aliases
LZP
Chromosome Location
Chr. 10: 72893556 - 72933036 on Build GRCh38
UniGene ID
Hs.8366
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152635.2 NP_689848.1 2 358
NR_130125.1 2 358
GenBank mRNA AK096435.1 - 2 333
AY013707.2 - 2 189
AY358339.1 - 2 311
BC126440.1 - 2 330 8992156
DA979981.1 - 2 358

View Full Product Details