Gene Transcripts

Gene Symbol
ARMCX4
Entrez Gene ID
100131755
Gene Name
armadillo repeat containing, X-linked 4
Gene Aliases
CXorf35, GASP4
Chromosome Location
Chr. X: 101418263 - 101535988 on Build GRCh38
UniGene ID
Hs.729062
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256155.2 NP_001243084.2 2 480
NR_028407.2 9 1100
NR_045861.1 6 991
NR_045864.1 6 1033
GenBank mRNA AK096955.1 - 6 789
AK126987.1 - 6 854
AK292543.1 - 6 824
BC032236.1 - 6 770
CR933662.1 - 9 1075

View Full Product Details