Gene Transcripts

Gene Symbol
SLC35G1
Entrez Gene ID
159371
Gene Name
solute carrier family 35 member G1
Gene Aliases
C10orf60, TMEM20
Chromosome Location
Chr. 10: 93893951 - 93914265 on Build GRCh38
UniGene ID
Hs.632085
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134658.1 NP_001128130.1 3 606
NM_153226.2 NP_694958.1 3 603
XM_011539371.2 XP_011537673.1 3 571
GenBank mRNA AK091309.1 - 3 603
BC041432.1 - 2 335
BC104814.1 - 3 577 8143817
BC143283.1 - 3 526 9051786
BC143284.1 - 3 577 9051787

View Full Product Details