Gene Transcripts

Gene Symbol
LRRC57
Entrez Gene ID
255252
Gene Name
leucine rich repeat containing 57
Gene Aliases
Chromosome Location
Chr. 15: 42528332 - 42548804 on Build GRCh38
UniGene ID
Hs.234681
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153260.2 NP_694992.2 5, 6 809
GenBank mRNA AK094131.1 - 5, 6 803
AK094891.1 - 4, 5 1043
AK309861.1 - 4, 5 1374
BC058935.1 - 5, 6 765 5578024
BX538324.1 - 4, 5 955
CN350663.1 - 2, 3 259
CR749223.1 - 5, 6 778

View Full Product Details