Gene Transcripts

Gene Symbol
RGL4
Entrez Gene ID
266747
Gene Name
ral guanine nucleotide dissociation stimulator like 4
Gene Aliases
Rgr
Chromosome Location
Chr. 22: 23690861 - 23699176 on Build GRCh38
UniGene ID
Hs.658997
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153615.1 NP_705843.1 1 1250
GenBank mRNA AY101396.1 - 1 1250

View Full Product Details