Gene Transcripts

Gene Symbol
WEE1
Entrez Gene ID
7465
Gene Name
WEE1 G2 checkpoint kinase
Gene Aliases
WEE1A, WEE1hu
Chromosome Location
Chr. 11: 9573681 - 9589767 on Build GRCh38
UniGene ID
Hs.249441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143976.1 NP_001137448.1 2 782
NM_003390.3 NP_003381.1 2 922
GenBank mRNA AK122837.1 - 2 782
BC051831.1 - 2 720 6147695
BC070052.1 - 2 912 30336707
BX641032.1 - 1 440
U10564.1 - 2 922
X62048.1 - 2 839

View Full Product Details