Gene Transcripts

Gene Symbol
TMEM86B
Entrez Gene ID
255043
Gene Name
transmembrane protein 86B
Gene Aliases
Chromosome Location
Chr. 19: 55226634 - 55229264 on Build GRCh38
UniGene ID
Hs.135215
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC023000.1 - 3 1607
BC033193.1 - 2 1369

View Full Product Details