Gene Transcripts

Gene Symbol
BMP7
Entrez Gene ID
655
Gene Name
bone morphogenetic protein 7
Gene Aliases
OP-1
Chromosome Location
Chr. 20: 57168753 - 57266651 on Build GRCh38
UniGene ID
Hs.473163
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001719.2 NP_001710.1 7 2066
GenBank mRNA AK094784.1 - 3 806
AK291186.1 - 7 1642
X51801.1 - 7 1659

View Full Product Details