Gene Transcripts

Gene Symbol
NDUFB1
Entrez Gene ID
4707
Gene Name
NADH:ubiquinone oxidoreductase subunit B1
Gene Aliases
CI-MNLL, CI-SGDH, MNLL
Chromosome Location
Chr. 14: 92116123 - 92121809 on Build GRCh38
UniGene ID
Hs.183435
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004545.3 NP_004536.2 3 341
GenBank mRNA AF050638.1 - 3 195
AK055348.1 - 3 2267
BC009691.1 - 3 201
BC104672.1 - 3 261
BC126232.1 - 3 338 8991948
BC126234.1 - 3 338 8991950
BM052981.1 - 3 341
BX476462.1 - 3 240

View Full Product Details