Gene Transcripts

Gene Symbol
PTPRH
Entrez Gene ID
5794
Gene Name
protein tyrosine phosphatase, receptor type H
Gene Aliases
R-PTP-H, SAP1
Chromosome Location
Chr. 19: 55181247 - 55209506 on Build GRCh38
UniGene ID
Hs.179770
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161440.2 NP_001154912.2 5 944
NM_002842.4 NP_002833.4 7 1478
XM_011527183.1 XP_011525485.1 7 1499
XM_011527188.1 XP_011525490.1 6 1448
XM_011527190.1 XP_011525492.1 7 1499
XM_017027056.1 XP_016882545.1 6 1486
XM_017027057.1 XP_016882546.1 6 1486
XM_017027058.1 XP_016882547.1 6 1519
XM_017027059.1 XP_016882548.1 6 1486
XM_017027060.1 XP_016882549.1 6 1486
XM_017027061.1 XP_016882550.1 4 952
XM_017027062.1 XP_016882551.1 6 1394
XM_017027063.1 XP_016882552.1 6 1387
XM_017027064.1 XP_016882553.1 6 1486
GenBank mRNA AK097257.1 - 7 1497
BC111715.1 - 6 894 40008777
BC111716.1 - 7 1428 40008775
D15049.2 - 7 1452

View Full Product Details