Gene Transcripts

Gene Symbol
PDZD9
Entrez Gene ID
255762
Gene Name
PDZ domain containing 9
Gene Aliases
C16orf65
Chromosome Location
Chr. 16: 21957540 - 22001137 on Build GRCh38
UniGene ID
Hs.98244
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173806.3 NP_776167.2 3 421
NR_033694.1 5 672
XM_006721029.3 XP_006721092.1 4 625
XM_011545785.2 XP_011544087.1 4 628
XM_017023109.1 XP_016878598.1 3 447
XM_017023110.1 XP_016878599.1 5 686
GenBank mRNA BC021724.1 - 5 677
BC028605.1 - 5 677 5265642
BC039562.1 - 5 669 5169242
DB450393.1 - 3 408

View Full Product Details