Gene Transcripts
- Gene Symbol
- P2RX1
- Entrez Gene ID
- 5023
- Gene Name
- purinergic receptor P2X 1
- Gene Aliases
- P2X1
- Chromosome Location
- Chr. 17: 3896591 - 3916666 on Build GRCh38
- UniGene ID
- Hs.41735
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002558.3 |
NP_002549.1
|
|
2857 |
|
GenBank mRNA |
X83688.1 |
- |
|
2590 |
|
View Full Product Details