Gene Transcripts

Gene Symbol
EMP3
Entrez Gene ID
2014
Gene Name
epithelial membrane protein 3
Gene Aliases
YMP
Chromosome Location
Chr. 19: 48325372 - 48330555 on Build GRCh38
UniGene ID
Hs.9999
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001313905.1 NP_001300834.1 3 181
NM_001425.2 NP_001416.1 3 365
XM_011526605.2 XP_011524907.1 4 296
GenBank mRNA AL541088.3 - 3 365
BC009718.1 - 3 172 3891050
BT007124.1 - 2 111
CR456871.1 - 2 111
CR541955.1 - 2 111
U52101.1 - 3 160
U87947.1 - 3 352
X94771.1 - 2 114

View Full Product Details