Gene Transcripts
- Gene Symbol
- RPL9
- Entrez Gene ID
- 6133
- Gene Name
- ribosomal protein L9
- Gene Aliases
- L9, NPC-A-16
- Chromosome Location
- Chr. 4: 39454124 - 39458948 on Build GRCh38
- UniGene ID
- Hs.412370
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000661.4 |
NP_000652.2
|
3, 4 |
226 |
|
NM_001024921.3 |
NP_001020092.1
|
3, 4 |
384 |
|
GenBank mRNA |
AB062431.1 |
- |
1 |
173 |
|
AK130966.1 |
- |
1 |
189 |
|
AK297178.1 |
- |
3, 4 |
209 |
|
AK303908.1 |
- |
1 |
189 |
|
AK312241.1 |
- |
1 |
193 |
|
AL039355.2 |
- |
3, 4 |
359 |
|
AY320412.1 |
- |
3, 4 |
209 |
|
AY376242.1 |
- |
2, 3 |
343 |
|
AY597415.1 |
- |
1 |
200 |
|
BC000483.2 |
- |
1 |
178 |
2964733 |
BC004156.2 |
- |
1 |
172 |
2961026 |
BC004206.2 |
- |
1 |
172 |
2961026 |
BC007967.2 |
- |
1 |
228 |
4304670 |
BC012149.1 |
- |
1 |
228 |
4554838 |
BC031906.1 |
- |
2, 3 |
173 |
4996986 |
BC066318.1 |
- |
1 |
193 |
5277491 |
BC070214.1 |
- |
1 |
232 |
6605055 |
BC100285.1 |
- |
2, 3 |
333 |
6022858 |
BG829769.1 |
- |
2, 3 |
174 |
|
BM841336.1 |
- |
3, 4 |
226 |
|
D14531.1 |
- |
2, 3 |
173 |
|
D16961.1 |
- |
1 |
146 |
|
U09953.1 |
- |
3, 4 |
196 |
|
U21138.1 |
- |
1 |
173 |
|
View Full Product Details