Gene Transcripts

Gene Symbol
RBMXL3
Entrez Gene ID
139804
Gene Name
RNA binding motif protein, X-linked like 3
Gene Aliases
CXorf55
Chromosome Location
Chr. X: 115189400 - 115192868 on Build GRCh38
UniGene ID
Hs.376008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145346.1 NP_001138818.1 1 1267
GenBank mRNA AK097568.1 - 1 680
AK301904.1 - 1 1267

View Full Product Details