Gene Transcripts

Gene Symbol
MRPS21
Entrez Gene ID
54460
Gene Name
mitochondrial ribosomal protein S21
Gene Aliases
MDS016, MRP-S21, RPMS21
Chromosome Location
Chr. 1: 150293842 - 150308979 on Build GRCh38
UniGene ID
Hs.405880
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018997.3 NP_061870.1 1, 2 616
NM_031901.5 NP_114107.1 2, 3 180
GenBank mRNA AB049955.1 - 2, 3 153
AF182417.1 - 1, 2 125
AK289467.1 - 2, 3 167
BC004566.2 - 2, 3 149 3633098
BC006554.2 - 1, 2 521 2819715
BI755132.1 - 1, 2 573
HY161691.1 - 2, 3 181
U79258.1 - 1, 2 537

View Full Product Details