Gene Transcripts
- Gene Symbol
- PODNL1
- Entrez Gene ID
- 79883
- Gene Name
- podocan like 1
- Gene Aliases
- SLRR5B
- Chromosome Location
- Chr. 19: 13931187 - 13953392 on Build GRCh38
- UniGene ID
- Hs.448497
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001146254.1 |
NP_001139726.1
|
2, 3 |
504 |
|
NM_001146255.1 |
NP_001139727.1
|
2, 3 |
504 |
|
NM_024825.3 |
NP_079101.3
|
2, 3 |
528 |
|
XM_006722901.1 |
XP_006722964.1
|
2, 3 |
461 |
|
XM_006722902.1 |
XP_006722965.1
|
2, 3 |
461 |
|
XM_006722903.1 |
XP_006722966.1
|
2, 3 |
461 |
|
XM_011528308.1 |
XP_011526610.1
|
2, 3 |
495 |
|
XM_011528309.2 |
XP_011526611.1
|
2, 3 |
474 |
|
XM_011528310.2 |
XP_011526612.1
|
2, 3 |
473 |
|
XM_011528311.1 |
XP_011526613.1
|
2, 3 |
461 |
|
XM_011528313.1 |
XP_011526615.1
|
2, 3 |
495 |
|
XM_011528314.1 |
XP_011526616.1
|
2, 3 |
461 |
|
XM_011528316.1 |
XP_011526618.1
|
2, 3 |
495 |
|
XM_017027313.1 |
XP_016882802.1
|
2, 3 |
495 |
|
GenBank mRNA |
AK027100.1 |
- |
2, 3 |
494 |
|
AK290731.1 |
- |
2, 3 |
461 |
|
AK296101.1 |
- |
1, 2 |
289 |
|
AK298489.1 |
- |
2, 3 |
504 |
|
BC057786.1 |
- |
2, 3 |
528 |
30347370 |
View Full Product Details