Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_053004.2 | NP_443730.1 | 8 | 1331 | |
GenBank mRNA | AB051432.1 | - | 8 | 1272 | |
AF238328.1 | - | 8 | 895 | ||
AF301895.1 | - | 8 | 1296 | ||
AY007378.1 | - | 8 | 1331 | ||
BC012060.1 | - | 8 | 1232 | 4548339 | |
CR456496.1 | - | 6 | 1128 |