Gene Transcripts

Gene Symbol
Glrx
Entrez Gene ID
64045
Gene Name
glutaredoxin (thioltransferase)
Gene Aliases
Glrx1, Grx
Chromosome Location
Chr. 2: 2606032 - 2615944 on Build Rnor_6.0
UniGene ID
Rn.1484
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022278.1 NP_071614.1 3 897
GenBank mRNA AF167981.3 - 3 897
BC061555.1 - 3 926 6889009
FQ224532.1 - 3 933

View Full Product Details