Gene Transcripts
- Gene Symbol
- Taar4
- Entrez Gene ID
- 294122
- Gene Name
- trace amine-associated receptor 4
- Gene Aliases
- Ta2, taR-2, taR-4
- Chromosome Location
- Chr. 1: 22558787 - 22559830 on Build Rnor_6.0
- UniGene ID
- Rn.138138
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175583.1 |
NP_783173.1
|
1 |
135 |
|
View Full Product Details