Gene Transcripts

Gene Symbol
Ucp1
Entrez Gene ID
24860
Gene Name
uncoupling protein 1 (mitochondrial, proton carrier)
Gene Aliases
Ucp, Ucpa, Uncp
Chromosome Location
Chr. 19: 24456976 - 24464808 on Build Rnor_6.0
UniGene ID
Rn.10281
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012682.2 NP_036814.1 2 397
GenBank mRNA BC088156.1 - 2 385
FM087156.1 - 2 396
M11814.1 - 2 395
X03894.1 - 2 307

View Full Product Details