Gene Transcripts

Gene Symbol
Sbp
Entrez Gene ID
25540
Gene Name
spermine binding protein
Gene Aliases
Chromosome Location
Chr. 10: 13289514 - 13309332 on Build Rnor_6.0
UniGene ID
Rn.9914
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008767547.1 XP_008765769.1 1 200
GenBank mRNA J02675.1 - 1 224

View Full Product Details