Gene Transcripts

Gene Symbol
Mndal
Entrez Gene ID
100040462
Gene Name
myeloid nuclear differentiation antigen like
Gene Aliases
Gm2785
Chromosome Location
Chr. 1: 173857220 - 173880260 on Build GRCm38
UniGene ID
Mm.463437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170853.1 NP_001164324.1 766
GenBank mRNA AK087931.1 - 4 595
AK136391.1 - 768
FJ594740.1 - 4 665
FJ594741.1 - 674

View Full Product Details