Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027892.2 | NP_082168.1 | 4 | 549 | |
GenBank mRNA | AK004785.1 | - | 4 | 549 | |
AK139570.1 | - | 4 | 434 | ||
BC125381.1 | - | 5 | 618 | 40129896 | |
BC137630.1 | - | 5 | 618 | 8860649 |