Gene Transcripts

Gene Symbol
Slc39a2
Entrez Gene ID
214922
Gene Name
solute carrier family 39 (zinc transporter), member 2
Gene Aliases
F730005G13Rik, Gm1789, Gm289, Zip2
Chromosome Location
Chr. 14: 51891972 - 51896745 on Build GRCm38
UniGene ID
Mm.281343
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039676.2 NP_001034765.2 4 1228
GenBank mRNA AK089322.1 - 4 1181
AK135117.1 - 3 2337
AK170178.1 - 5 794
BC113769.1 - 4 1198 40057862

View Full Product Details