Gene Transcripts

Gene Symbol
Ttc37
Entrez Gene ID
218343
Gene Name
tetratricopeptide repeat domain 37
Gene Aliases
mKIAA0372
Chromosome Location
Chr. 13: 76098730 - 76190323 on Build GRCm38
UniGene ID
Mm.277964
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081352.1 NP_001074821.1 12 1224
GenBank mRNA AK134255.1 - 12 1170
EF591876.1 - 836

View Full Product Details