Gene Transcripts

Gene Symbol
Zbtb16
Entrez Gene ID
235320
Gene Name
zinc finger and BTB domain containing 16
Gene Aliases
AI467657, PLZF, Zfp145, lu
Chromosome Location
Chr. 9: 48651797 - 48836381 on Build GRCm38
UniGene ID
Mm.457803
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033324.2 NP_001028496.1 7 4227
GenBank mRNA AK142947.1 - 7 4228

View Full Product Details