Gene Transcripts

Gene Symbol
Atr
Entrez Gene ID
245000
Gene Name
ataxia telangiectasia and Rad3 related
Gene Aliases
Chromosome Location
Chr. 9: 95857597 - 95952760 on Build GRCm38
UniGene ID
Mm.212462
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019864.1 NP_063917.1 34 5878
GenBank mRNA AK132223.1 - 8 1003
AK220176.1 - 6 684

View Full Product Details