Gene Transcripts

Gene Symbol
Slc35g2
Entrez Gene ID
245020
Gene Name
solute carrier family 35, member G2
Gene Aliases
Gm519, Tmem22
Chromosome Location
Chr. 9: 100552188 - 100571085 on Build GRCm38
UniGene ID
Mm.236289
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101483.1 NP_001094953.1 2 357
GenBank mRNA BY726562.1 - 2 357
CB588129.1 - 2 363

View Full Product Details