Gene Transcripts

Gene Symbol
Nbea
Entrez Gene ID
26422
Gene Name
neurobeachin
Gene Aliases
Lyst2, mKIAA1544
Chromosome Location
Chr. 3: 55625195 - 56183701 on Build GRCm38
UniGene ID
Mm.384353
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030595.1 NP_085098.1 14, 15 2526
GenBank mRNA Y18276.1 - 14, 15 2526

View Full Product Details